Human Diseases Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T240 T240-15
470353-188BX 81.5 CAD
470353-188 470353-042
Human Diseases Microslides
Slides Prepared Slides
Learn about the organisms and conditions that cause various human ailments.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects It on both sides
  • Set of 15 Includes detailed lesson pan on the topic under study
  • Durable, inexpensive teaching resource
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set Includes: 1. Viral diseases - Influenze virus (320,000×) 2. Bacterial diseases - Tuberculosis (100×) and (1000×) 3. Protis diseases - Amoebic dysentary (100×) 4. Fungal diseases - Candida Albicans 5. Parasitic diseases - Trichina worm (65×) 6. Genetic diseases - Sickle cell anemia (400×) 7. Environmental illnesses - normal (150×), Blackened lung (65×) 8. Multifactorial diseases - Atherosclerosis (3×) and 15×).