Human AC3 ELISA Kit
Supplier: ANTIBODIES.COM LLC
A314276-96
77175-470EA 877.69 CAD
77175-470
Human AC3 ELISA Kit
Assays ELISAs
Human AC3 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AC3 in serum, plasma, and other biological fluids.
  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human AC3 ELISA kit (A314276) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human AC3 in serum, plasma or other biological fluids. An antibody specific for AC3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the AC3 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-AC3 antibody, which also binds the AC3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-AC3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-AC3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of AC3 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of AC3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.