The Microscope - A Tool for Learning Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T207 T207-15
470353-190EA 19.51 CAD
470353-190 470353-176
The Microscope - A Tool for Learning Microslides
Slides Prepared Slides
Explore how the microscope allows us to see important details of life and the world around us.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. The letter 'E' (100×) 2. Cheek cells (100 and 400×) 3. Cheek cells (900×) 4. Onion skin cells (110×) 5. Hydra (50×) and (175×) 6. Paramecium (200×) 7. Paramecium - interference microscope (200×) 8. Paramecium (1500×).