Life in the Sea Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T126 T126-15
470353-182BX 83.1 CAD
470353-182 470353-100
Life in the Sea Microslides
Slides Prepared Slides
Explore the many forms of ocean life.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Diatoms (220×) 2. Asterionella (200×) 3. Zooplankton - Radiolarians (30×) 4. Zooplankton - Zoea larva (25×) 5. Sponge spicules (30×) 6. Jellyfish tentacle - C.S. (100×) 7. Starfish (1×) 8. Shark scales (13×).