How a One-Celled Animal Divides
Supplier: SUPERTEK SCIENTIFIC LLC
T104 T104-15
470353-158EA 19.51 CAD
470353-158 470353-146
How a One-Celled Animal Divides
Slides Prepared Slides
Learn about how unicellular organisms divide.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Paramecia - fresh (10×) 2. Paramecium - fresh -(100×) 3. Paramecium - stained (100×) 4. Division - stage 1 - stained (100×) 5. Stage 2 - stained - (100×) 6. Stage 3 - stained - (100×) 7. Stage 4 - stained - (100×) 8. Paramecium - all stages - stained (10×).