From Tadpole to Frog Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T016 T016-15
470353-064EA 19.51 CAD
470353-064 470353-140
From Tadpole to Frog Microslides
Slides Prepared Slides
Learn about how the frog develops from tadpole stage to an adult.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Frog's egg 2. Frog embryo 3. Tadpole - early stage 4. Tadpole - outside gills 5. Tadpole - inside gills 6. Young frog 7. Young frog-tail disappearing 8. Adult frog.