Asexual Reproduction Microslides
Supplier: SUPERTEK SCIENTIFIC LLC
T063 T063-15
470353-172EA 19.51 CAD
470353-172 470353-128
Asexual Reproduction Microslides
Slides Prepared Slides
Explore the key steps of asexual reproduction.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
  • Set of 15 includes detailed lesson pan on the topic under study
  • Durable, economical teaching resource
Introduce junior and senior high school students to the world of microscopy with economical, unbreakable microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides.
Each microslide is accompanied by a detailed lesson plan designed to stimulate, inform, and question students about the topic under study. Each lesson plan has a pocket in which the microslide is stored. Lesson plans are printed on heavy card stock and will serve for repeated use.
Ordering information: Viewers are available separately.
Delivery information: Set includes 1. Binary fission-Paramecium, Budding-yeast (900×) 3. Budding-hydra (25×) 4. Sporulation-yeast (1200×) 5. Sporulation-bread mold (10×) 6. Sporulation-fern (300×) 7. Regeneration-planarian (5×) 8. Vegetative propagation.