Mineral Oil Dropper Bottle
Supplier: Agilent
FS-SMO15 FS-SMO15
CA76645-242EA 83.55 CAD
76645-242 CA76645-242
Mineral Oil Dropper Bottle
Lubricants
Used as an overlay on DNA samples to minimize evaporation.