Picus® 2 Multi-Channel Pipettes, Electronic
Supplier: Sartorius
LH-747421 LH-747441 LH-747461 LH-747491 LH-747321 LH-747341 LH-747361 LH-747391
CA77200-826EA 3094.69 CAD
CA77200-826 CA77200-828 CA77200-830 CA77200-832 CA77200-818 CA77200-820 CA77200-822 CA77200-824
Picus® 2 Multi-Channel Pipettes, Electronic
Pipettes Micropipettes
Picus® 2 pipettes offer reliable, ergonomic pipetting for all users. They feature customizable programs, safety features, and connect to the Sartorius app for automatic adjustments and improved workflow, enhancing laboratory productivity.
  • Reliable and repeatable pipetting results
  • Wide selection of pipetting modes
  • Customizable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app
The Picus® 2 pipettes ensure reliable and repeatable pipetting results with unbeatable ergonomic design that is kind to your hand. Picus® 2 is the tool of choice for everybody from a student to experienced laboratory professional. It is as intuitive to use as a mechanical pipette and yet offers a broad range of options even for the most advanced users.
Picus® 2 saves you time in the laboratory with a wide selection of pipetting modes and customizable programs for every need. The most frequently used modes with specified settings can be saved on your device for a quick and easy access whereas safety features like password protection and calibration reminders will enhance your compliance. By connecting Picus® 2 to your mobile device, you can use the Sartorius pipetting mobile app to smoothly run sample preparation workflows and adjust the pipette setting automatically taking your productivity to the next level.
Sterility: Non sterile
Type of battery: Lithium polymer
Medical device: No medical devices