Scrubber for DK and DKL Series of Kjeldahl Digestion Units, KS 1000
Supplier: VELP Scientifica
F307A0660
77257-076EA 8697.09 CAD
77257-076
Scrubber for DK and DKL Series of Kjeldahl Digestion Units, KS 1000
Digestion Systems Mineralisation Workstations
Designed to neutralize toxic fumes produced during mineralization and safeguard laboratory operators without requiring any connection to tap water.
  • Safeguard laboratory environment and operators
  • Neutralizes corrosive toxic gases
  • Quick and easy cleaning and maintenance
The KS 1000 Scrubber effectively neutralizes corrosive and toxic gases in two steps:
Dilution: In the right tank, reaction gases flow into a water solution, where they are effectively dissolved. This step allows even part of the sulphur dioxide produced during digestion to be trapped.
Neutralization: In the left glass tank, reaction gases are neutralised in a sodium hydroxide solution. Thanks to a wide contact surface between gas and liquid, the KS 1000 Scrubber achieves maximum neutralization efficiency.