ZymoBIOMICS 96 MagBead DNA Kit
Supplier: Zymo Research
D4306-E D4308-E D4302-E
77020-296EA 1156.62 CAD
77020-296 77020-266 77020-326
ZymoBIOMICS 96 MagBead DNA Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Inhibitor-free microbial DNA from any sample validated for microbiome and metagenomic analyses.
  • High-throughput purification of high quality, inhibitor-free DNA from any sample including feces, soil, water, biofilms, swabs, saliva, and body fluids
The ZymoBIOMICS DNA kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenome analyses. The ZymoBIOMICS lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. gram-negative/positive bacteria, fungus, protozoans, and algae), making it ideal for microbial community profiling. Uniform mechanical lysis of all microbes is achieved by bead beating with the innovative ultra-high density bashingbeads. This kit is equipped with our OneStep PCR inhibitor removal technology, enabling PCR amplification from DNA derived from inhibitor-rich environmental samples. Purified DNA is ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing. DNA Size is 15-20 kb.