Large Volume Conical Centrifuge Tubes
Supplier: MTC BIO, INC.
C0175 C0225
470353-208CS 385.5 CAD
470353-208 470353-204
Large Volume Conical Centrifuge Tubes
Tubes Centrifuge Tubes
These polypropylene tubes are durable and centrifuge compatible.
  • Molded graduations
  • Leakproof screw caps
  • Sterile
  • RNase, DNase, and Pyrogen-free
Large volume conical centrifuge tubes offer a maximum RCF of 6000×g. These tubes may be used within a temperature range of –80 to 120 °C.