Eisco Test Tube Peg Racks
Supplier: Wards
CH0711B2 CH0711B1
470344-302EA 27.8 CAD
470344-302 470344-378
Eisco Test Tube Peg Racks
Racks Tube Racks
Plastic peg rack provides convenient insertion and removal of inverted test tubes.
  • Peg racks with a sturdy plastic construction that will stand up to the wear and tear of the lab
  • Supports up to 50 or 96 inverted test tubes that measure up to 13 or 16 mm in diameter
  • Features holes through the base to increase air flow and facilitate drying
  • Rack can also be used to hold and dry electrophoresis and chromatography plates
These plastic peg racks conveniently hold test tubes in an inverted position to facilitate drying while minimizing airborne contaminants. The bottom of the rack has holes to increase air flow. In addition to test tubes, the drying rack can be used to hold and dry electrophoresis and chromatography plates. Diameter of pegs measure 4 mm.