Eisco Glass Stirring Rods with Spade End
Supplier: Wards
CH0755A CH0755B
470344-018EA 9.49 CAD
470344-018 470345-276
Eisco Glass Stirring Rods with Spade End
Stirrers Stirring Rods
Glass rods with one spade end and one button end for stirring, scooping, and crushing.
  • Pack of ten multipurpose stirring rods with one spade end and one button end
  • Made of high quality, heavy-duty borosilicate 3.3 glass - can withstand high temperatures and thermal shocks
  • Great for stirring, scooping, circulating and crushing
Multipurpose stirring rods with one spade end and one button end. Great for stirring, scooping, circulating and crushing. Each rod measures 6 mm in diameter. The spade end of the rod measures approximately 15 mm long and wide, and the button end measures approximately 10 mm in diameter. Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco Labs borosilicate glass has the following heat tolerances:
Strain point: 515 °C
Annealing point: 565 °C
Softening point: 820 °C