RightCare KN95 Protective 5-Ply Face Masks (Non-Medical)
Supplier: OUTSOURCE PHARMACY SOLUTIONS, INC.
12889-001-4 12889-001-4CASE
77148-064PK 12.89 CAD
77148-064 77148-066
RightCare KN95 Protective 5-Ply Face Masks (Non-Medical)
Masks Hygiene Masks
RightCare premium KN95 disposable face masks provide superior protection while maintaining easy breathability and a comfortable fit. With five layers of protection, they are designed with a fluid repellent non-woven outer layer, a highly efficient interior triple layer filtration system, and a soft, absorbent non-woven inner layer.
  • Masks provide an effective physical barrier to help prevent microscopic particles from entering
  • Highly efficient filtration from a triple-layer filtration system
  • Filtration efficiency is greater than 95%
  • Fluid-resistant non-woven material safely guards against blood, water, and other airborne fluids
  • Disposable KN95 masks are packaged in re-sealable bags of ten
  • Comfortable fit with earloops and a shapeable nose bridge wire
Layers of protection offer excellent general use protection, comfort, and breathability. A triple-layer filtration system ensures easy breathing while keeping unwanted particles out. Intended for every day use at work, school, travel, or shopping, these masks provide a filtration efficiency greater than 95%.
Fluid-resistant non-woven outer layer serves as the first line of defense to protect the user and the inner filter elements of the mask. It safely guards against blood, water, and other airborne fluids. Comfortable earloops and a shapeable nose bridge wire ensures a snug fit over the face. The disposable and lightweight design makes the mask easy to use during travel, work, shopping, and other activities.
Masks are disposable and cannot be washed. Store in a dry and ventilated location. This KN95 mask is not to be used as a replacement for gas masks or as a medical mask or worn in high-risk contamination environments.
Packaging: Masks are packaged 10 per bag, 100 bags per case.