InterMask Disposable Premium 3-Ply Face Masks, ASTM Level 3
Supplier: OUTSOURCE PHARMACY SOLUTIONS, INC.
12888-001BLACK 12888-001D 12888-001D40
77148-056BX 13.7 CAD
77148-056 77148-058 77148-060 77148-062
InterMask Disposable Premium 3-Ply Face Masks, ASTM Level 3
Masks Hygiene Masks
Single pleated three ply rectangular face mask with a shapeable nosepiece and two elastic ear loops to hold the mask firmly in place. ASTM level 3 disposable masks feature a hydrophobic non-woven outer layer, a meltblown middle filtration layer, and a soft absorbent non-woven inner layer.
  • High quality pleasted facemasks tested to ASTM level 3 at Nelson labs
  • Three layers of protection help filter microbes from entering and exiting mask, while maintaining breathability
  • Soft and comfortable fabric with strong elastic ear loops; one size fits all
  • Adjustable and easy to use with a shapeable nosepiece that contours to any face
Masks provide 160 mm Hg fluid resistance, greater than 98% BFE (Bacterial Filtration Efficiency), and greater than 98% PFE (Particle Filtration Efficiency) at 0.1 µm. These protective face masks are made from high-quality, non-woven fabric, which offers increased filtration of fluids in air compared to woven cloth. Elastic ear strap loops are strong, rounded for comfort, and slip easily over the ears. These lightweight pleated facemasks are made with a shapeable nosepiece that contours to any face and creates a light seal. The non-woven fabric allows for effortless breathing while in use. Replace mask every two hours for best protection to ensure moisture from breath doesn't dampen mask.
The exterior hydrophobic non-woven layer prevents fluids from entering the mask. The middle meltblown filtration layer provides high-level static filtration, effectively preventing micro dust and contaminants from entering. The inner layer fabric is made of a soft absorbent non-woven fabric that will not irritate the face.
Certifications: Tested to ASTM level 3 at Nelson labs.
Packaging: Masks are packaged 50 per box, 40 boxes per case.