Amino Labs Bioplastic Kits: Make a Bioplastic Bag™
Supplier: AMINO LABS NORTH INCORPORATED SE
WWG017-G WWG017-S
470352-668KT 312.55 CAD
470352-668 470352-656
Amino Labs Bioplastic Kits: Make a Bioplastic Bag™
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Construct your own green destiny!
  • Develop some basic skills for making bioplastics including mixing and extruding liquid plastics
  • Become familiar with common bioplastic ingredients—their names, the sources from which they derive, and their purpose in the creation of bioplastics
  • Learn the fundamental principles of the biodegradation process with the optional extension activity
In just three 30-minute sessions, any beginners can learn how to make a bioplastic bag using the Bioplastic Kit: Make a Bioplastic Bag™.
This kit provides all the necessaryMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable