RightCare No-Touch Infrared Forehead Thermometers
Supplier: OUTSOURCE PHARMACY SOLUTIONS, INC.
12894-005-3 12894-005-35CT
77148-070EA 48.35 CAD
77148-070 77148-072
RightCare No-Touch Infrared Forehead Thermometers
Every family, office, business, and school needs a safe, accurate, and reliable thermometer. The RightCare digital non-contact Infrared Thermometer for adults and children is designed using the latest infrared technology. It includes the newest generation of core chip sensing that provides accurate readings within ±0.3 °C in under 0.5 seconds, without skin contact.
  • Non-contact sensor allows for safe, non-invasive body temperature readings at least 2" from forehead
  • Instant accurate readings in Fahrenheit or Celsius display on LCD screen within 0.5 seconds
  • Multifunctional design measures body, room, or surface temperature
  • Color display and audible tone make it easy to identify safe temperatures
  • 32 sets of data storage
  • Two AA batteries included
  • Limited 18 months manufacturer warranty
Easy to use with adults and children. New 2020 generation sensor is designed with an advanced algorithm that provides faster, more accurate results, and allows for safe, non invasive body temperature readings at least 2" from the forehead. Readings in Fahrenheit or Celsius display on the large, easy-to-read, back-lit LCD screen within 0.5 seconds. The infrared sensor and fast microprocessor can detect thermal heat on bodies and surfaces.
Designed for use by healthcare professionals or consumers. It's perfect for any doctor's office, clinic, hospital, school, workplace, and household. The unit is equipped with three modes: Body, Room, and Surface Temperature. A color display and audible tone makes it easy to identify safe temperatures. Green indicates normal, orange indicates a slight fever, and red with an audible tone indicates a high fever. The audible tone setting can be turned off if desired. 32 sets of data storage allow users to save and keep track of temperatures with the click of a button. Includes enough storage room to easily record and track changes in family members temperature as well.