Eisco® 3-Prong Universal Support Clamps, Large
Supplier: Wards
CH0684E CH0684C CH0684D
470343-670EA 28.39 CAD
470343-670 470344-492 470345-112
Eisco® 3-Prong Universal Support Clamps, Large
Clamps General Purpose Clamps
3-prong retort support clamps have angled jaws that open to hold items up to 100 mm diameter.
  • 3-Prong Support Clamp made of diecast alloy
  • Holds items up to 100 mm in diameter
  • Angled jaw design ensures a first grip at all angles of opening
  • Overal length is 27 cm
The 3-Prong Support Clamp is ideal for clamping glassware of various sizes onto retort rods. Available with PVC coating or cork lining on the jaws to protect vessels from being scratched. Adjustable jaw capacity is 0 to 100 mm.