Eisco® 3-Prong Support Clamps, PVC Coated
Supplier: Wards
CH0684 CH0684A
470344-012EA 21.35 CAD
470344-012 470345-024
Eisco® 3-Prong Support Clamps, PVC Coated
Clamps General Purpose Clamps
3-prong retort support clamps with angled, PVC coated jaws that open to hold items up to 90 mm in diameter.
  • 3-Prong Support Clamp made of diecast alloy
  • Holds items up to 90 mm in diameter
  • PVC coated jaws protect glass from scratches
  • Angled jaw design ensures a first grip at all angles of opening
  • Overall length is 25 cm
The 3-Prong Support Clamp is ideal for clamping glassware of various sizes onto retort rods. The PVC coating on the jaws protects vessels from being scratched. Clamp has a 5.65" long rod which is 8mm in diameter and will fit into most bossheads. Adjustable jaw capacity is 0 to 90 mm.