Eisco® 4-Prong Support Clamps, Cork Lined
Supplier: Wards
CH0682 CH0682A
470344-988EA 21.35 CAD
470344-988 470345-602
Eisco® 4-Prong Support Clamps, Cork Lined
Clamps General Purpose Clamps
Retort support clamps with four cork-lined prongs and curved jaws for a secure grip.
  • 4-prong support clamp made of diecast alloy and plated brass
  • Holds items up to 90 mm in diameter
  • Cork-lined jaws protect glass from scratches
  • Curved jaw design ensures a first grip at all angles of opening
  • Overall length is 25 cm
The 4-Prong Support Clamp is ideal for clamping glassware of various sizes onto retort rods. The cork lining on the jaws protects vessels from being scratched. Clamp has a 5.65" long rod which is 8mm in diameter and will fit into most bossheads. Adjustable jaw capacity is 0 to 90 mm.