C-Clamps, Eisco
Supplier: Wards
CH0938C CH0938E
470345-378EA 47.85 CAD
470345-378 470345-632
C-Clamps, Eisco
Clamps General Purpose Clamps
Steel C-Clamps with threaded screw and swivel pads for a gentle yet secure hold.
  • C-Clamps made of drop-forged steel
  • Threaded steel screw with swivel pads grips material more tightly
  • Sliding T-bar handle allows for easy tightening and untightening
  • Use for lab apparatus setup, woodworking, fabrication, and even for lessons on pressure and torque
  • Great in the lab or in the shop
C-Clamp for securing objects against flat surfaces, like lab benches. The drop-forged steel construction is built to last. The threaded screw is topped with a swivel pad for safer, more secure clamping.