Ward's® Safety Bottle Carriers
Supplier: Heathrow Scientific
120912W 120913W 120914W 120915W 120916W 120917W 120918W 120911W
470352-502CS 597.25 CAD
470352-502 470352-500 470352-504 470352-490 470352-492 470352-498 470352-494 470352-496
Ward's® Safety Bottle Carriers
Carriers Bottle Carriers/Containers
Secure and convenient bottle transport.
  • Robust design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors, with designated labeling areas for easy identification
  • Made of chemical-resistant thermoplastic elastomer/PP blend
The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable