PIP® AR/FR Dual Certified Coveralls with Insect Repellent
Supplier: PIP
9100-2110D/2X 9100-2110D/3X 9100-2110D/4X 9100-2110D/5X 9100-2110D/L 9100-2110D/M 9100-2110D/S 9100-2110D/XL
76945-016EA 225.47 CAD
76945-016 76945-018 76945-020 76945-022 76945-012 76945-010 76945-008 76945-014
PIP® AR/FR Dual Certified Coveralls with Insect Repellent
Workwear
Coveralls are manufactured from a cotton/nylon blend to provide comfort and durability, with a built-in insect repellent for additional personal protection.
  • ATPV 8 cal/cm²
  • PPE category: 2
  • 7 oz. woven FR fabric (90% cotton/10% nylon)
  • Aramid high tenacity FR thread
  • No metal pieces on the garment
  • Bar-tacked at all stress points
Designed for use in electrical and utility applications, these coveralls feature a protective stand-up collar with hook and loop closure, storm guard with hook and loop closure, and a comfort vented back for breathability and increased mobility. The garments have a side elasticized waist, FR rib knit cuffs, and two waist patch pockets with side access.
Certifications: NFPA 1975-2009, NFPA 2112-2012 ,NFPA 70E 2012. ASTM F 1506-2010a. CAN/CGSB 155.20-2000, CAN/CGSB 155.22-97. OSHA 29 CFR1910.269. EN ISO 11611:2008, EN ISO 11612:2008, EN ISO 14116:2008. UL model # STC24.