PIP® AR/FR Dual-Certified Coveralls with Vented Back
Supplier: PIP
9100-2100D/5X 9100-2100D/L 9100-2100D/M 9100-2100D/S 9100-2100D/XL 9100-2120D/XL 9100-2100D/2X 9100-2100D/3X 9100-2100D/4X 9100-2120D/2X 9100-2120D/3X 9100-2120D/4X 9100-2120D/5X 9100-2120D/L 9100-2120D/M 9100-2120D/S
76945-006EA 357.85 CAD
76945-006 76944-996 76944-994 76944-992 76944-998 76945-030 76945-000 76945-002 76945-004 76945-032 76945-034 76945-036 76945-038 76945-028 76945-026 76945-024
PIP® AR/FR Dual-Certified Coveralls with Vented Back
Workwear
Coveralls are manufactured from a cotton/nylon blend to provide comfort and durability.
  • ATPV 8 cal/cm²
  • PPE category: 2
  • 7 oz. woven FR fabric (90% cotton/10% nylon)
  • Aramid high tenacity FR thread
  • No metal pieces on the garment
  • Bar-tacked at all stress points
Designed for use in electrical and utility applications, these coveralls feature a protective stand-up collar with hook and loop closure, storm guard with hook and loop closure, and a comfort vented back for breathability and increased mobility. The garments have a side elasticized waist, FR rib knit cuffs, and two waist patch pockets with side access.
Certifications: UL certified. NFPA 1975-2009, NFPA 2112-2012, NFPA 70E 2012. ASTM F 1506-2010a. CAN/CGSB 155.20-2000, CAN/CGSB 155.22-97. OSHA 29 CFR1910.269. EN ISO 11611:2008, EN ISO 11612:2008, EN ISO 14116:2008.