Eisco Rectangular Retort Bases with Rubber Strips, Steel
Supplier: Wards
CH0649A CH0649B CH0649C
470344-592EA 28.1 CAD
470344-592 470344-818 470345-624
Eisco Rectangular Retort Bases with Rubber Strips, Steel
Retort Stands
Premium Retort Bases with no-slip rubber strips and tapped hole made of powder-coated pressed steel.
  • Rectangular retort bases made of corrosion resistant, powder coated pressed steel
  • Rubber strips on sort ends prevent slipping and protect surfaces
  • Sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-weighted Retort Stands are made of corrosion resistant, powder coated pressed steel, and feature rubber strips at the short ends prevent slipping and protect surfaces. With a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.