Eisco Triangular Retort Bases
Supplier: Wards
CH0648A CH0648B CH0648C
470343-870EA 42.05 CAD
470343-870 470345-684 470345-954
Eisco Triangular Retort Bases
Retort Stands
Cast iron Triangular Retort Stand with center-tapped hole for maximum stability in minimum footprint.
  • Three-point retort bases made of chemical-resistant cast iron with a black coating
  • Offers maximum stability in a minimal footprint
  • Sturdy construction provides superb support for experimental setups
  • Features a centered-tapped hole for a 10×1.5 mm threaded rod (not included)
Triangular 3-Point Retort Stands are made of chemically-resistant cast iron with a black finish. Features a centered tapped hole suitable for rods with a 10×1.5 mm metric thread.More Product Information