Eisco T-Shaped Nesting Retort Bases, Cast Iron
Supplier: Wards
CH0656G CH0656H
470343-606EA 48.8 CAD
470343-606 470344-976
Eisco T-Shaped Nesting Retort Bases, Cast Iron
Retort Stands
T-Shaped Retort Bases designed to nest closely to each other, made of chemically-resistant cast iron.
  • T-shaped retort bases made of chemically-resistant cast iron with a black coating
  • Designed with one raised leg that allow multipe bases to slide freely under one another for compact setup or storage
  • Heavy and sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
T-Shaped Retort Stands are made of chemically-resistant cast iron with a black coating. One raised leg allows multipe bases to slide freely under one another for compact setup or storage. Features a tapped hole approximately ¹/₃ of the way from the intersection of the legs, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for both science classrooms and professional labs.