Eisco Two-Hole Rectangular Retort Bases, Steel
Supplier: Wards
CH0656C CH0656B
470344-024EA 35.1 CAD
470344-024 470345-122
Eisco Two-Hole Rectangular Retort Bases, Steel
Retort Stands
Red steel Retort Bases with two tapped holes support ring stand, titration, or specific gravity setups.
  • Rectangular retort bases made of corrosion resistant, powder coated pressed steel
  • Features two tapped holes - one centered and one at end - for 10×1.5 mm threaded rods (not included)
  • Ideal for supporting standard ring stands or titration and specific gravity setups
  • Sturdy construction provides superb stability for experimental setups
  • Red color
Red-colored Retort Stands are made of corrosion resistant, powder coated pressed steel. They feature two tapped rod holes - one centered and one at a short side end - suitable for rods with 10×1.5 mm metric threads. Provides superb stability for standard ring stands or titration and specific gravity setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.