Eisco Heavy-Duty Rectangular Retort Bases, Cast Iron
Supplier: Wards
CH0654A CH0654B
470345-982EA 66.75 CAD
470345-982 470346-048
Eisco Heavy-Duty Rectangular Retort Bases, Cast Iron
Retort Stands
Low profile, heavy-duty Retort Bases made of chemically-resistant cast iron with a smooth coating.
  • Rectangular retort bases made of chemical-resistant cast iron with smooth, fine-coated finished
  • Low, 1" tall profile
  • Heavy and sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Heavy-duty Retort Stands are made of chemically-resistant cast iron and have a fine, smooth coating. Features a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for both science classrooms and professional labs.