Eisco Rectangular Retort Bases, Steel
Supplier: Wards
CH0652A CH0652C CH0652B CH0652D
470343-794EA 17.05 CAD
470343-794 470344-886 470345-144 470345-608
Eisco Rectangular Retort Bases, Steel
Retort Stands
Economical Retort Bases with tapped rod hole made of powder-coated pressed steel.
  • Rectangular retort bases made of corrosion resistant, powder coated pressed steel
  • Sturdy construction provides superb stability for experimental setups
  • Features a tapped hole for a 10×1.5 mm threaded rod (not included)
Retort Stands are made of corrosion resistant, powder coated pressed steel, and feature a tapped hole at one end, suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for science classrooms.