TETPOR AIR DEMI Capsules
Supplier: PARKER HANNIFIN TE
ZEMTA-020GG-AN3 ZEMTA-020HH-AN3 ZEMTA-020TT-AN3 ZEMTB-020GG-AN3 ZEMTB-020HH-AN3 ZEMTB-020TT-AN3 ZEMTE-020GG-AN3 ZEMTE-020HH-AN3 ZEMTE-020TT-AN3
76529-530PK 1795.51 CAD
76529-530 76529-532 76529-534 76529-536 76529-538 76529-540 76529-542 76529-544 76529-546
TETPOR AIR DEMI Capsules
Filters Membrane Filter Capsules
TETPOR AIR sterilization filter capsules offer exceptional filtration performance while providing the highest levels of biosecurity throughout the process industry.
  • Assured biosecurity with absolute rated filtration
  • High flow rates with low pressure drops
  • High voids volume PTFE membrane
  • Unique prefilter layer
  • Silicone seal material
Operating at ambient temperature conditions, TETPOR AIR filter capsules provide a cost-effective filtration solution. A polypropylene prefilter layer extends service life in heavily contaminated environments.
TETPOR AIR filter capsules also utilize a well-proven, inherently hydrophobic expanded PTFE membrane validated as sterilizing grade in liquid in accordance with ASTM F838-05. This ensures the removal of all airborne bacteria, viruses and bacteriophage.