QRP® Qualakote® Nylon Gloves
Supplier: PIP
PDESDEC-2X PDESDEC-2XS PDESDECL PDESDECM PDESDECS PDESDECXL PDESDECXS
76945-126CS 559.46 CAD
76945-126 76945-114 76945-122 76945-120 76945-118 76945-124 76945-116
QRP® Qualakote® Nylon Gloves
Gloves
Seamless knit nylon glove with polyurethane coated Microfoam grip on the palm and fingertips.
  • Seamless nylon knit shell is comfortable and reduces hand fatigue
  • Knit wrist prevents dirt and debris from entering the glove
  • Polyurethane coating provides a secure grip in dry and slightly wet/oily conditions
  • Low-lint gloves knit from nylon filament yarns
  • Superior comfort, dexterity, and tactile sensitivity
  • Nylon yarns absorb perspiration and leave hands cool and comfortable
  • Reusable and launderable
Ideal for product handling, finishing and assembly,More Product Information