Parker® PROPOR HC DEMI Capsules
Supplier: PARKER HANNIFIN TE
ZEHCA-620GG-PN3 ZEHCA-620GG-PS3 ZEHCA-620HH-PN3 ZEHCA-620TT-PN3 ZEHCA-620TT-PS3 ZEHCB-620GG-PN3 ZEHCB-620GG-PS3 ZEHCB-620HH-PN3 ZEHCB-620HH-PS3 ZEHCB-620TT-PN3 ZEHCB-620TT-PS3 ZEHCE-620GG-PN3 ZEHCE-620GG-PS3 ZEHCE-620HH-PN3 ZEHCE-620HH-PS3 ZEHCE-620TT-PN3 ZEHCE-620TT-PS3 ZEHCA-620HH-PS3
76529-496PK 2086.5 CAD
76529-496 76529-498 76529-500 76529-502 76529-504 76529-506 76529-508 76529-510 76529-512 76529-514 76529-516 76529-518 76529-520 76529-522 76529-524 76529-526 76529-528 76554-140
Parker® PROPOR HC DEMI Capsules
Filters Membrane Filter Capsules
PROPOR HC sterilizing grade filters have been specifically designed for the effective and economical processing of difficult to filter solutions.
  • Optimized membrane configuration allows up to 10× the throughput compared to single layer membrane products
  • Integral prefilter layer can condense filter trains for greater processing economy
  • Incorporates a fully validated and integrity testable 0.2 µm membrane for assurance of sterility
  • Low binding for minimal product loss
  • Single-use capsule format available with pre-sterile version by gamma-irradiation
The optimized PROPOR HC PES membrane configuration features a highly asymmetric membrane prefilter layer, which significantly extends throughput and prevents the problems associated with premature filter blockage with complex solutions. PROPOR HC filters are high capacity and fast flowing.
The PES membrane is inherently low binding, which minimizes product loss due to protein or preservative adsorption. The filters have low extractable levels and broad chemical compatibility.
Seals material: Silicone.