ActivGrip™ Nitrile Coated Chemical Resistant Gloves
Supplier: PIP
56-AG585/XXL 56-AG586/L 56-AG586/M 56-AG586/S 56-AG586/XL 56-AG586/XS
76944-986DZ 113.54 CAD
76944-986 76944-982 76944-980 76944-978 76944-984 76944-976
ActivGrip™ Nitrile Coated Chemical Resistant Gloves
Gloves
Nitrile coated gloves with a cotton liner and MicroFinish™ grip, available in 10" or 12" lengths.
  • Revolutionary surface treatment provides superior grip in dry, wet and oily conditions
  • MicroFinish™ grip adjusts to varied conditions by reacting like tiny suction cups
  • Seamless cotton liner for comfort and improved fit
  • Tremendous comfort and dexterity
Ideal for general assembly of oily parts, metal handling, finishing and machine operation. The MicroFinish™ coating provides excellent resistance to liquid permeation, while maintaining strong protection against punctures and abrasion.
Packaging: Gloves are packaged 6 dozen per case.