Assurance® Unsupported Nitrile Chemical Resistant Gloves
Supplier: PIP
50-N2272G/L 50-N2272G/M 50-N2272G/XL 50-N2272G/XXL
76944-970DZ 158.71 CAD
76944-970 76944-968 76944-972 76944-974
Assurance® Unsupported Nitrile Chemical Resistant Gloves
Gloves
Gloves provides superior resistance to physical hazards such as cut, puncture, snag, and abrasion, as well as many chemical hazards including solvents and oil based chemicals.
  • Unsupported nitrile with sandpatch grip
  • Unlined to reduce potential of product contamination
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Chlorinated to harden and cleanse the surface of the glove
  • Silicone-free
Ideal for chemical processing, oil refining and petrochemicals, food processing, aerospace and automotive degreasing, automotive assembly and painting, machining operations, metal fabrication, graphic arts and printing cleanup, furniture manufacturing and electronics.
Certifications: Complies with FDA food handling requirements 21 CFR, Part 177.
Packaging: Gloves are packaged 1 dozen per case.