Eisco Plastic Lab Pitchers
Supplier: Wards
ECH0139H ECH0139G ECH0139B ECH0139E ECH0139C ECH0139D ECH0139A ECH0139F ECH0139SET
470343-594EA 60.2 CAD
470343-594 470343-728 470343-896 470344-192 470344-202 470344-632 470345-704 470345-898 470346-086
Eisco Plastic Lab Pitchers
Beakers
Pitchers/beakers with screen printed graduations made from chemical-resistant polypropylene plastic.
  • Made of high quality, heat and chemical resistant polypropylene
  • Short form with easy to read, screen printed graduations and specifications
  • Transparent - excellent optical quality
  • Features a non-drip spout
  • Autoclavable and microwavable
Made of high quality, heavy duty polypropylene. Short form. Beakers feature a non-drip spout. Transparent, with easy to read screen printed graduations and specifications. Autoclavable and microwavable with superior chemical durability.