APM Adult Body Bag, 16 mil PVC Laminated Polyester, Center/Envelope Zipper, 6 Handles, Red
Supplier: Mopec
BE801-R-2XL BE801-R-3XL BE801-R-4XL BE801-R-XL BE801E-R-2XL BE801E-R-3XL BE801E-R-4XL BE801E-R-XL BE801-R BE801E-R
76945-278CS 1528.76 CAD
76945-278 76945-280 76945-282 76945-284 76945-258 76945-260 76945-262 76945-264 76945-276 76945-256
APM Adult Body Bag, 16 mil PVC Laminated Polyester, Center/Envelope Zipper, 6 Handles, Red
Bags Storage Bags
Mopec in partnership with American Professional Manufacturing provide excellent body bags.
  • Custom-made, lockable slider for easy opening and closing
  • HD nylon zipper, 100% sewn with nylon thread
  • Dielectrically sealed seams
  • Available with center or envelope zipper
  • Six handles secured by heavy-duty steel rivets
  • Leak resistance
Materials: 16 mil PVC laminated polyester
Dimensions: Various sizes (Available in Bariatric XL, 2XL, 3XL and 4XL).