Eisco Woulff Gas Wash Bottles
Supplier: Wards
CH0511B CH0511C CH0511D CH0511A
470344-086EA 75.75 CAD
470344-086 470344-238 470344-512 470345-616
Eisco Woulff Gas Wash Bottles
Bottles Gas Washing Bottles
Woulff gas wash bottles with two necks for preparing gases.
  • Made of mechanically strong and chemical resistant Borosilicate 3.3 glass
  • Two glass necks with ground sockets
  • Used to remove impurities from gases, trapping particles and droplets, and for pressure equalization
  • Autoclavable at 121 °C
Woulff gas wash bottles made from high quality, heavy duty Borosilicate 3.3 glass. Ground glass joints allow for easy sealing.