Eisco Cylindrical Gas Jars
Supplier: Wards
CH0494E CH0494B CH0494C CH0494F CH0494D CH0494A
470343-780EA 28.4 CAD
470343-780 470344-092 470344-358 470345-498 470345-634 470345-732
Eisco Cylindrical Gas Jars
Jars General Use Jars
Gas jars made of soda lime glass ideal for collecting gases in a closed system.
  • Made of thick soda-lime glass
  • Used to collect gases during lab experiments or to examine living specimens
  • Not for heating applications
  • Use watch glasses as lids (not included)
Gas jar is made of soda-lime glass and, with its broad base and wide mouth, is ideal for collecting gases generated during various science experiments. Watch glasses are typically used as lids and are available separately.