Blank for Calibration Kit, A30
Supplier: FireFlySci
A30
76016-668EA 317.56 CAD
76016-668
Blank for Calibration Kit, A30
Racks Cuvette Racks
Blank for use in UV/VIS calibration kits.
  • Anodized aluminum construction
  • External dimensions: 12.5L×12.5W×45H mm