Sub-Micro Cuvette with Threaded PTFE Stopper (Capacity 10 to 160 µl), Type 701M
Supplier: FIREFLYSCI, INC.
701MUV10.10B 701MUV10.10C 701MUV10.50A 701MUV10.50B 701MUV10.50C 701MUV10.100A 701MUV10.100B 701MUV10.100C 701MUV10.160A 701MUV10.160B 701MUV10.160C
76016-114EA 1792.04 CAD
76016-114 76016-116 76016-118 76016-526 76016-528 76016-530 76016-532 76016-534 76016-536 76016-538 76016-540
Sub-Micro Cuvette with Threaded PTFE Stopper (Capacity 10 to 160 µl), Type 701M
Cuvettes Spectrophotometer Cuvettes
FireflySci's 10mm pathlength sub-micro spectrophotometer cuvette with capacities ranging from 10 to 160 µl.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in UV Quartz
  • Available in 10mm lightpath
  • Capacities of 10, 50, 100, 160 µl
  • Two windows optically polished
  • Includes PTFE threaded stopper
Great for DNA, RNA, and protein micro samples.