SHURMount™ Plus Liquid Coverslip
Supplier: General Data
SHURMount™ Plus
SMP-4 SMP-4
CA76632-838CS 185.3 CAD
76632-838 CA76632-838
SHURMount™ Plus Liquid Coverslip
Cover Glasses
Spreads thinly and evenly to form a permanent, scratch-resistant layer that acts as a coverslip or can be used in conjunction with a coverslip.
  • Toluene based, compatible with most applications
  • Minimizes the need to purchase coverslips
Acrylic resin allows rapid drying for hand coverslipping.