Control Slides, Normal Human Tissue, Medullated Nerve
Supplier: Mopec
SC933-100 SC933-25
76948-170CS 2509.28 CAD
76948-170 76948-172
Control Slides, Normal Human Tissue, Medullated Nerve
Slides Microscopy Control Slides
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µ
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 6More Product Information