ReadiLink™ iFluor® 488 Oligo and ssDNA Labeling Kit
Supplier: AAT BIOQUEST INC
17480
17481
76779-094EA
437.22
CAD
76779-094
76779-076
ReadiLink™ iFluor® 488 Oligo and ssDNA Labeling Kit
Protein Linking and Labeling Systems
The kit is optimized for efficient labeling and contains all the essential reagents required for efficient labeling of ssDNA or oligos.
- Efficient labelling
- Contains all essential reagents
- Multiple applications
The resulting iFluor® 488-labeled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.