ReadiLink™ DIG (Digoxigenin) Oligo and ssDNA Labeling Kit
Supplier: AAT Bioquest
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Efficient labelling
- Contains all essential reagents
- Multiple applications
The resulting DIG-labeled oligo and ssDNA probes are ideally suited for a variety of biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.