ReadiLink™ Cy5 Oligo and ssDNA Labeling Kit
Supplier: AAT BIOQUEST INC
17488
17489
76779-114EA
437.22
CAD
76779-114
76779-022
ReadiLink™ Cy5 Oligo and ssDNA Labeling Kit
Protein Linking and Labeling Systems
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
- Robust protocol
- Bright labeling
The resulting Cy5-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.