ReadiLink™ iFluor® 647 Oligo and ssDNA Labeling Kit
Supplier: AAT BIOQUEST INC
17484 17485
76779-060EA 437.22 CAD
76779-060 76779-102
ReadiLink™ iFluor® 647 Oligo and ssDNA Labeling Kit
Protein Linking and Labeling Systems
The kit is optimized for efficient labelling and contains all the essential reagents required for efficient labelling of ssDNA or oligos.
  • Robust protocol
  • Bright labeling
  • Photostable fluorescence
The resulting iFluor® 647-labelled DNA probes are ideally suited for biological applications, e.g., electrophoretic mobility shift assays (EMSA), Northern and Southern blots, colony or in situ hybridizations.
Packaging: Packaged in plastic vials.
Caution: For research use only.