ReadiLink™ iFluor® 555 Nick Translation dsDNA Labeling Kit
Supplier: AAT BIOQUEST INC
17402 17403
76779-028EA 665.53 CAD
76779-028 76779-106
ReadiLink™ iFluor® 555 Nick Translation dsDNA Labeling Kit
Protein Linking and Labeling Systems
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
  • Provides all necessary reagents
  • Adjustable ratios
  • Wide sample type compatibility
The resulted iFluor® 555-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridization (FISH).
Packaging: Packaged in plastic vials.
Caution: For research use only.