ReadiLink™ iFluor® 488 Nick Translation dsDNA Labeling Kit
Supplier: AAT BIOQUEST INC
17400
17401
76779-074EA
665.53
CAD
76779-074
76779-044
ReadiLink™ iFluor® 488 Nick Translation dsDNA Labeling Kit
Protein Linking and Labeling Systems
It is compatible with a wide variety of sample materials, including bacterial artificial chromosome (BAC) DNA, human genomic DNA, purified PCR products, supercoiled and linearized plasmid DNA.
- Provides all necessary reagents
- Adjustable ratios
- Wide sample type compatibility
The resulted iFluor® 488-labeled DNAs can be used in a variety of molecular biology techniques such as fluorescence in situ hybridization (FISH).
Packaging: Packaged in plastic vials.
Caution: For research use only.